Find methods for your needs
"Green" and ultrafast method for determination of soft drink ingredients on the Thermo Scientific Acclaim 120 C18 RSLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of soft drink ingredients. The separation is performed on a Thermo Scientific Acclaim 120 C18 RSLC column in under 1.5 minutes. To avoid the use of toxic solvents, acetonitrile has been replaced by ethanol. To reduce the solvent viscosity and column pressure, the column is heated to 50 °C. Wavelength switching (changes from 230 nm to 214 nm at 0.3 minutes) suppresses interferences near acesulfame-K.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, Acesulfame-K, Aspartame, Aspartylphenylalanine, Benzoic acid, Caffeine, HPLC, Sodium saccharin, Soft drink, Sorbate potassium, UltiMate 3000 RSLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.