Find methods for your needs
Improved analysis of water-soluble vitamin standards using a Thermo Scientific Acclaim C30 column
Description
The Thermo Scientific Dionex UltiMate 3000 RS system is applied for the analysis of seven water-soluble vitamins. The separation was performed in less than 12 minutes on a Thermo Scientific Acclaim C30 5 µm column, with UV detection at 210 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim C30, Ascorbic acid, Folic acid, HPLC, Niacinamide, Pantothenic acid, Pyridoxine, Riboflavin, Thiamine, Water-soluble vitamin |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.