Find methods for your needs
Improved isocratic separation of basic, neutral, and acidic molecules on a Thermo Scientific Acclaim Mixed-Mode WAX-1 column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used for the isocratic separation of basic, neutral and acidic pharmaceuticals.The novel column chemistry of the Acclaim Mixed-Mode WAX-1 column provides a multi-mode separation mechanism that includes hydrophobic, anion-exchange and cation-exclusion interactions.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WAX-1, Acidic, Basic, HPLC, Isocratic, Neutral, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.