Find methods for your needs
Fast analysis of ingredients in a toothache pain-reliever using a Thermo Scientific Acclaim Mixed-Mode WAX-1 column
Description
The Thermo Scientific Acclaim Mixed-Mode WAX-1 column is used for the separation of ingredients in a toothache pain-reliever, with UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Acclaim Mixed-Mode WAX-1, Benzocaine, HPLC, Saccharin, Sorbate, Toothache pain-reliever, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.