Find methods for your needs
Direct Analysis of Multicomponent Adjuvants by HPLC with Charged Aerosol Detection
Description
The Thermo Scientific Dionex Ultimate 3000 LC system with Corona Veo RS Charged Aerosol Detector quantifies adjuvant components including saponins, lipids and detergents after separation on a Thermo Scientific Hypersil GOLD PFP 1.9 µm column.Market: | BioPharma |
Keywords: | Biotherapeutics, Glycoside, Lipid, Saponin, Universal Detection |
Matrix: | Adjuvant formulation in deionized water, Standards in aqueous solvent |
For Research Use Only. Not for use in diagnostic procedures.