Find methods for your needs
Simultaneous determination of penicillin G and its potassium counterion
Description
The Thermo Scientific Acclaim Trinity P2 provides baseline separation of both penicillin G and potassium ion with excellent resolution and good peak shape using a simple isocratic method.| Market: | Pharma |
| Keywords: | Counterion, HPLC, Penicillin G, Charged aerosol detection, Potassium |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.