Find methods for your needs
An improved fat-soluble vitamin separation using a Thermo Scientific Acclaim RSLC PolarAdvantage HPLC column
Description
The Thermo Scientific Ultimate 3000 system is applied for the improved separation of fat-soluble vitamins. The Thermo Scientific Acclaim RSLC PolarAdvantage column separated nine vitamins with a run time of less than 20 minutes.| Market: | Food and Beverage |
| Keywords: | Acclaim RSLC PolarAdvantage, Fat soluble vitamins, HPLC, Retinol acetate, Tocopherol, UV detection, Vitamin D2, Vitamin D3, Vitamin E acetate, Vitamin K1, Vitamin K2 |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.