Find methods for your needs
High resolution analysis of the major benzalkonium homologs in nasal spray using HPLC-CAD
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis benzalkonium in nasal spray. The separation is performed using a Thermo Scientific Acclaim Surfactant Plus column, with charged aerosol detection.| Market: | Pharma |
| Keywords: | Benzalkonium, Surfactant, Acclaim Surfactant Plus, Charged aerosol detection, CAD, Nasal spray |
| Matrix: | Nasal spray |
For Research Use Only. Not for use in diagnostic procedures.