Find methods for your needs
Rapid organic acid analysis using the Thermo Scientific Acclaim Surfactant Plus HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the fast analysis of vitamin C and its decomposition products. The separation is performed isocratically on a Thermo Scientific Acclaim Surfactant Plus, with UV detection at 220 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim Surfactant Plus, ascorbate, HPLC, Organic Acids, Oxalate, Sorbate, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.