Find methods for your needs
Improved analysis of fenoprofen according to the USP monograph using a Thermo Scientific Accucore XL C8 column
Description
The Thermo Scientific HPLC system is applied for the analysis of fenoprofen according to the USP monograph. The separation is performed on a Thermo Scientific Accucore XL C8 HPLC column, with UV detection at 272 nm.| Market: | Pharma |
| Keywords: | Accucore XL C8, Fenoprofen, Gemfibrozil, HPLC, USP monograph, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.