Find methods for your needs
Analysis of fenoprofen using the USP method with a Thermo Scientific Syncronis C8 HPLC column
Description
The USP monograph for Fenoprofen, requires an L7 column with 5µm packing, therefore a Thermo Scientific Syncronis C8, 5µm column would be suitable and is demonstrated in this work.| Market: | Pharma |
| Keywords: | Fenoprofen, Gemfibrozil, HPLC, Syncronis C8, USP, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.