Find methods for your needs
Fast analysis of cephalosporins using a Thermo Scientific Syncronis aQ HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the determination of cephalosporins. The separation is performed on a Thermo Scientific Syncronis C18 HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Cefazolin, Cephalexin, Cephaloridine, Cephalosporins, Cephradine, HPLC, Syncronis C18, UltiMate 3000, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.