Find methods for your needs
Improved LC-MS/MS analysis of Nicotine, Cotinine and Trans-3-hydroxycotinine in urine using Thermo Scientific SOLA SCX SPE and a Thermo Scientific Syncronis C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of nicotine, cotinine and trans-3-hydroxycotinine in urine. The samples were prepared using a SOLA SCX solid phase extraction (SPE) 96 well plate, and the separation performed in under 6 minutes on a Thermo Scientific Syncronis C18 HPLC column, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Cotinine, LC-MS/MS, MS detection, Nicotine, SOLA CX, SPE, Trans-3-hydroxycotinine, Urine |
| Matrix: | Urine |
For Research Use Only. Not for use in diagnostic procedures.