Find methods for your needs
Fast dextromethorphan and dextrorphan analysis with excellent peak shape using a Thermo Scientific Accucore C18 HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the determination of dextromethorphan and dextrorphan. The separation is performed on a Thermo Scientific Accucore C18 HPLC column, with UV detection at 194 nm.| Market: | BioPharma |
| Keywords: | Accucore C18, Dextromethorphan, Dextrorphan, HPLC, UltiMate 3000, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.