Find methods for your needs
Rapid LC-MS/MS determination of HCTZ and Losartan from human plasma using Thermo Scientific SOLA SCX SPE and a Thermo Scientific Accucore aQ HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of Losartan and HCTZ in human plasma. The samples were prepared using SOLA SCX solid phase extraction (SPE), and the separation performed in 2 minutes on a Thermo Scientific Accucore aQ HPLC column, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore aQ, Diuretic, Furosemide, HCTZ, LC-MS/MS, Losartan, MS detection, SOLA CX, SPE |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.