Find methods for your needs
Fast analysis of non-steroidal anti-inflamatory drugs (NSAIDs) using a Thermo Scientific Syncronis C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of non-steroidal anti-inflamatory drugs (NSAIDs). The separation is performed on a Thermo Scientific Syncronis C18 HPLC column, with UV detection at 260 nm.| Market: | Pharma |
| Keywords: | Aspirin, Diclofenac, Fenoprofen, HPLC, Ibuprofen, Naproxen, NSAID, Piroxicam, Sulindac, Syncronis C18, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.