Find methods for your needs
Fast analysis of benzodiazepines using a Thermo Scientific Hypersil GOLD Phenyl UHPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of benzodiazepines. The separation is performed on a Thermo Scientific Hypersil GOLD Phenyl 1.9 µm UHPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Acetominophen, benzodiazepines, Diazepam, Di-isopropylphthalate, Di-n-propylphthalate, HPLC, Hydroxybenzoic acid, Hypersil GOLD Phenyl, Oxazepam, Uracil, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.