Find methods for your needs
Refine by Feature
More...
More...
More...
Name
More...
CAS Number
More...
Class
More...
Manufacturer
More...
Model
More...
Stationary Phase
More...
Dimensions
Length
Diameter/Tubing Size ID
Tubing Size OD
Particle Size/Film Thickness
Displaying 1-5 of 2542 results
Items per page:
Sort by
TN003773 Configuring a combustion-ion chromatography system using a complete workflow
Instrument Type: IC
A new C-IC total workflow system has been introduced: the Thermo Scientific™ Cindion™ Combustion/Absorption Module, and an autosampler that can be configured for solid or liquid samples. A standalone Thermo Scientific™ Cindion™ LPG/gas Module is also an option. In this technical note, we describe the configuration and operation of the Cindion C-IC system with the following configurations: Cindion combustion/absorption Module with Cindion autosampler: 1. Solids sampler and with the offline Cindion adsorption module needed for U.S. EPA Method 1621. 2) Solids sampler, 3.Liquids sampler.
AN 002998: Determination of selenite and selenate in environmental waters by IC-MS
Instrument Type: ICMS
To develop a method to determine selenite and selenate in spiked environmental water by coupling ion chromatography (IC) with single quadrupole mass spectrometry
AP003822: Screening of PFAS in food contact materials: Utilizing a new combustion-IC system for total organic fluorine (TOF) analysis
Instrument Type: IC
In AN003644, we developed a method to measure TOF in FCM using combustion-ion chromatography (C-IC). This application proof note demonstrates the determination of TOF in FCM using an enhanced C-IC system. The Thermo Scientific™ Cindion™ C-IC System combines the Thermo Scientific™ Dionex™ Inuvion™ IC System with the Thermo Scientific™ Cindion™ Combustion/Absorption Module, which minimizes PFAS contamination sources and includes a compact Z-fold combustion tube alongside full system control via Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) software.
Case study: Instrument and LC column migration for the purification and analysis of synthetic oligonucleotides
Instrument Type: HPLC (Biocompatible)
Application note 003192 - A simple method to purify oligonucleotides via the Thermo Scientific™ Vanquish™ Analytical Purification LC System and to perform quality control using UHPLC-UV
AN003774: Enhanced screening of PFAS compounds in wastewater: Implementing U.S. EPA Method 1621 with improved combustion-ion chromatography
Instrument Type: IC
Per- and polyfluoroalkyl substances (PFAS) is the collective name for over 14,000 synthetic fluorinated compounds. In addition to their extensive presence, PFAS compounds are persistent and bioaccumulate. Consequently, PFAS compounds are an environmental contamination concern. Toxicological studies of several PFAS compounds indicate the potential for acute to chronic effects impacting reproductive health. In this application, EPA Method 1621 is implemented on the Thermo Scientific Cindion C-IC designed for more efficient combustions and to minimize contamination sources.