Find methods for your needs
AP003822: Screening of PFAS in food contact materials: Utilizing a new combustion-IC system for total organic fluorine (TOF) analysis Be the first to rate this application
Description
In AN003644, we developed a method to measure TOF in FCM using combustion-ion chromatography (C-IC). This application proof note demonstrates the determination of TOF in FCM using an enhanced C-IC system. The Thermo Scientific™ Cindion™ C-IC System combines the Thermo Scientific™ Dionex™ Inuvion™ IC System with the Thermo Scientific™ Cindion™ Combustion/Absorption Module, which minimizes PFAS contamination sources and includes a compact Z-fold combustion tube alongside full system control via Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) software.
Market: Material Science
Keywords: pyrolysis, Dionex Inuvion IC system, Dionex IonPac AS24 column, Cindion combustion/absorption module
Matrix: food contact material
Author: Jingli Hu, Neil Rumachik
Affiliation: Thermo Fisher Scientific
Uploaded on 5/19/2025.
For Research Use Only. Not for use in diagnostic procedures.