Find methods for your needs
TN003773 Configuring a combustion-ion chromatography system using a complete workflow
Description
A new C-IC total workflow system has been introduced: the Thermo Scientific™ Cindion™ Combustion/Absorption Module, and an autosampler that can be configured for solid or liquid samples. A standalone Thermo Scientific™ Cindion™ LPG/gas Module is also an option. In this technical note, we describe the configuration and operation of the Cindion C-IC system with the following configurations: Cindion combustion/absorption Module with Cindion autosampler: 1. Solids sampler and with the offline Cindion adsorption module needed for U.S. EPA Method 1621. 2) Solids sampler, 3.Liquids sampler.| Market: | Environmental; Metals & Mining; Electronics & Semiconductors; Food and Beverage; Petrochem, Oil and Gas |
| Keywords: | halide, iodide, fluoride, Chloride, Sulfate, Bromide, PFAS, pyrolysis, total sulfur, total chlorine, total fluorine, C-IC, Cindion, combustion-IC, EPA 1621, total bromine, total iodine, Cindion Adsorption module, Cindion parts |
| Matrix: | Wastewater |
| Author: | Terri Christison and Neil Rumachik |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.