Find methods for your needs
AN003774: Enhanced screening of PFAS compounds in wastewater: Implementing U.S. EPA Method 1621 with improved combustion-ion chromatography Be the first to rate this application
pfas wastewater improved cic
Description
Per- and polyfluoroalkyl substances (PFAS) is the collective name for over 14,000 synthetic fluorinated compounds. In addition to their extensive presence, PFAS compounds are persistent and bioaccumulate. Consequently, PFAS compounds are an environmental contamination concern. Toxicological studies of several PFAS compounds indicate the potential for acute to chronic effects impacting reproductive health. In this application, EPA Method 1621 is implemented on the Thermo Scientific Cindion C-IC designed for more efficient combustions and to minimize contamination sources.
Market: Environmental
Keywords: Dionex IonPac AS24, fluoride, Fluorine, IonPac AS24, PFAS, total fluorine, Inuvion, AOF, Perfluoroalkyl substances, EPA Method 1621, adsorbable organic fluorine, C-IC, Cindion, Cindion C-IC, polyfluroalkyl substances, TF, combustion-IC
Matrix: Surface water, Wastewater, Water
Author: Terri Christison and Neil Rumachik
Affiliation: Thermo Fisher Scientific
Uploaded on 4/17/2025.
For Research Use Only. Not for use in diagnostic procedures.