Find methods for your needs
Rapid high temperature analysis of uracil and it's metabolites using a Thermo Scientific Hypercarb HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the determination of uracil and it's metabolites. The separation is performed at 150 °C on a Thermo Scientific Hypercarb high temperature HPLC column, with UV detection at 220 nm.
Market: Pharma
Keywords: Dihydrouracil, Fluorouracil, High temperature, HPLC, Hypercarb, Uracil, UV detection
Matrix: N/A
Uploaded on 8/29/2012.
For Research Use Only. Not for use in diagnostic procedures.