Find methods for your needs
Rapid high temperature analysis of uracil and it's metabolites using a Thermo Scientific Hypercarb HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of uracil and it's metabolites. The separation is performed at 150 °C on a Thermo Scientific Hypercarb high temperature HPLC column, with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | Dihydrouracil, Fluorouracil, High temperature, HPLC, Hypercarb, Uracil, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.