Find methods for your needs
A demonstration of time saving and recovery improvements for the extraction of beta blockers using Thermo Scientific HyperSep SLE compared to liquid / liquid extraction
Description
Thermo Scientific HyperSep solid-supported liquid-liquid extraction (SLE) is compared to traditional liquid-liquid extraction (LLE) for the extraction of beta blockers. The analysis is performed on a Thermo Scientific HPLC system using a Thermo Scientific Accucore C18 HPLC column. By using Thermo Scientific HyperSep SLE instead of traditional LLE methods, recovery of the analytes can be improved and the extraction time significantly reduced. For Research Use Only. Not for use in diagnostic procedures.| Market: | Clinical Research |
| Keywords: | Accucore C18, Beta blockers, HPLC, HyperSep SLE, Liquid-liquid extraction, LLE, Metoprolol, Pindolol, Plasma, Propranolol, Solid-supported liquid-liquid extraction, UV detection |
| Matrix: | Plasma |
For Research Use Only. Not for use in diagnostic procedures.