Description
Five-fold increase in method throughput compared to original method (fifty samples/hour). Associated 10-fold reduction in cost per sample through reduced mobile phase consumption and waste generation. Additional reduced method complexity from easy-to-prepare mobile phase| Market: | Pharma |
| Keywords: | Accucore C8, Fenoprofen, Gemfibrozil, Pharmaceutical, Solid core, USP, QA/QC, Drug Development, USP-NF Chapter 621, Small molecule, Vanquish Flex UHPLC, USP Modernization |
| Matrix: | Standard |
| Author: | Derek Hillbeck |
| Affiliation: | Thermo Fisher Scientific, Chromatography Consumables, Runcorn, UK |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.