Find methods for your needs
Rapid LC-MS/MS method for the analysis of five aritifical sweeteners using a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of five artificial sweeteners. The separation was performed on a Thermo Scientific Accucore RP-MS HPLC column in under one minute, with MS/MS detection.| Market: | Food and Beverage |
| Keywords: | Accucore RP-MS, Acesulfame potassium, Aspartame, LC-MS/MS, MS detection, Saccharin, Sodium cyclamate, Sucralose, Sweeteners |
| Matrix: | Standard |
For Research Use Only. Not for use in diagnostic procedures.