Find methods for your needs
Improved analysis of intact proteins using a Thermo Scientific Accucore 150-C4 150 Å pore diameter nanoLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of intact proteins (*indicates an impurity from carbonic anhydrase). The separation is performed on a Thermo Scientific Accucore 150-C4 150 Å pore diameter nanoLC column, with UV detection at 214 nm. The 150 Å pore size enables the effective analysis of molecules unable to penetrate into smaller diameter pores, whilst the low hydrophobicity C4 phase results in protein separation by hydrophobicity.| Market: | BioPharma |
| Keywords: | Accucore 150-C4, Carbonic anhydrase, Cytochrome C, Insulin, Intact protein, Myoglobin, nanoLC, Ovalbumin, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.