Find methods for your needs
Improved analysis of Cola soft drink additives using a Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of Cola soft drink additives. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column in less than 8 minutes, with UV detection at 254 nm. Cola soft drink is a complex matrix and as limited sample preparation has been performed in this analysis some matrix peaks are being detected.| Market: | Food and Beverage |
| Keywords: | Aspartame, Benzoic acid, Caffeine, Cola, HPLC, Hypersil GOLD, Saccharin, Soft drink additivies, UV detection |
| Matrix: | Cola soft drink |
For Research Use Only. Not for use in diagnostic procedures.