Find methods for your needs
Determination of Chloride and Sulfate in Crude Oil by Combustion Ion Chromatography Be the first to rate this application
Description
In this application, crude oil was combusted in the Mitsubishi AQF-2100H system and then analyzed using a Dionex Integrion Ion Chromatography system to determine chloride and sulfate with a Dionex IonPac AS28-4µm anion exchange column. In the chromatogram shown, the chloride concentration was ~17 ppm and sulfate was ~3000 ppm.
Market: Petrochem, Oil and Gas
Keywords: RFIC, CIC, Combustion, Corrosive, Integrion HPIC, Pyrohydrolysis, TN72211, TN000767
Matrix: Crude oil
Author: Carl Fisher
Affiliation: Thermo Fisher Scientific
Uploaded on 1/20/2017.