Find methods for your needs
Determination of Chloride and Sulfate in Crude Oil by Combustion Ion Chromatography
Description
In this application, crude oil was combusted in the Mitsubishi AQF-2100H system and then analyzed using a Dionex Integrion Ion Chromatography system to determine chloride and sulfate with a Dionex IonPac AS28-4µm anion exchange column. In the chromatogram shown, the chloride concentration was ~17 ppm and sulfate was ~3000 ppm.| Market: | Petrochem, Oil and Gas |
| Keywords: | RFIC, CIC, Combustion, Corrosive, Integrion HPIC, Pyrohydrolysis, TN72211, TN000767 |
| Matrix: | Crude oil |
| Author: | Carl Fisher |
| Affiliation: | Thermo Fisher Scientific |