Find methods for your needs
Ultrafast hydrophilic interaction UHPLC analysis of 2AA labelled N-glycans from monoclonal antibodies (mAbs)
Description
The Thermo Scientific Vanquish UHPLC system with Fluorescence detection is applied for the rapid glyco-profiling of a commercial chimeric IgG1 mAb.The separation is performed on a Thermo Scientific Accucore-150-Amide HILIC column in under 2.5 minutes. The developed methodology offers an excellent analytical platform for ultrafast glycan profile comparison of different batches of these mAbs.| Market: | BioPharma |
| Keywords: | Biotherapeutics, Glycosylation, IgG, N-glycans, Rituximab, Glycoprofiling, Monoclonal Antibodies (mAbs),, Accucore-Amide-HILIC, NIBRT, Bioprocessing Research |
| Matrix: | Rituximab drug product 10 mg/mL (MabThera®) |
| Author: | Silvia Millán, Stefan Mittermayr, Amy Farrell and Jonathan Bones |
| Affiliation: | NIBRT, Dublin, Ireland |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.