Find methods for your needs
Fast and Reproducible peptide mapping of an Antibody Drug Conjugate (Kadcyla) by using a SMART digest Kit
Description
The Thermo Scientific Dionex Ultimate 3000 Bio-LC system is applied for the peptide mapping analysis of an Antibody Drug Conjugate (Kadcyla). The separation was performed on a Thermo Scientific Acclaim RSLC 120-C18 column by using a Thermo Scientific SMART digest kit at UV detection of 214 nm. With a run time of 55 minutes the method allows separation of all peptides of Kadcyla (Antibody Drug Conjugate). The SMART Digest kit provides significant improvements in reproducibility, which results in fewer sample failures, higher throughput and the ability to more easily interrogate data.| Market: | BioPharma |
| Keywords: | Peptide mapping, UV detection, UHPLC, Antibody Drug Conjugate, Kadcyla, Trypsin digestion, SMART Digest kit |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.