 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
AN1158: HPAE-PAD Determination of Carbohydrates in Honey to Evaluate Samples for Quality and Adulteration
Description
We developed a HPAE-PAD method to assay fructose, glucose, and to measure the entire profile of di- and trisaccharides in honey. The separation was achieved on the CarboPac™ PA210-4μm column. Carbohydrate detection was by PAD with a gold working electrode and therefore no sample derivatization was required. In this work, honey sugars of 12 honey samples have been characterized and quantitated by HPAE-PAD. In addition, HPAE-PAD profiling was demonstrated as a method to study the adulteration of honey samples with commercial sugar syrups. An ICS-6000 can be used for this application.| Market: | Food and Beverage | 
| Keywords: | Carbohydrates, ICS-5000+, Honey, CarboPac PA210-4 μm column, food adulteration, sugar syrup, honey sugars authenticity, ICS-6000, TN 20, TN 70671, TN20, TN70671, AN 1158 | 
| Matrix: | Honey | 
| Author: | Manali Aggrawal and Jeff Rohrer | 
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA | 
For Research Use Only. Not for use in diagnostic procedures.