Find methods for your needs
Determination of Halogens and Sulfate in Liquid Petroleum Gas Using Combustion Ion Chromatography
Description
In this application, Liquid Petroleum Gas (LPG) samples were combusted in the Mitsubishi AQF-2100H system and then fluoride, chloride, and sulfate were determined using the Dionex ICS-2100 Ion Chromatography system with a Dionex IonPac AS15 anion-exchange column set.| Market: | Petrochem, Oil and Gas |
| Keywords: | IonPac AS15, RFIC, CIC, Combustion, Corrosive, AQF-2100H, halogens, LPG, Propane, Butane, Oxidative pyrohydrolysis, TN000767 |
| Matrix: | LPG |
| Author: | Carl Fisher |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.