Find methods for your needs
Analysis of Immunosupressant Drugs in Whole Blood Using a High-Efficiency Method on an Affordable High-Resolution Mass Spectrometer
Description
Clinical research labs commonly use selective and cost-efficient LC-MS techniques for the analysis of immunosupressants drugs. Usually, the quantitative method is developed on a triple quadrupole mass spectrometer. Here, we evaluated a high-efficiency method implemented on a Q Exactive Focus hybrid quadrupole Orbitrap mass spectrometer for improved selectivity when compared to the conventional solution.| Market: | Clinical Research |
| Keywords: | Clinical Research, Immunosuppressant drugs, Q Exactive Focus, tacrolimus, cyclosporin A, sirolimus, everolimus |
| Matrix: | Blood |
| Author: | Mindy Gao, Marta Kozak |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.