Find methods for your needs
Fast method for the determination of beta blockers from urine using Thermo Scientific SOLA CX SPE and a Thermo Scientific Accucore C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of beta blockers from urine. The samples are prepared using Thermo Scientific SOLA CX solid phase extraction (SPE) cartridges, and the separation is performed on a Thermo Scientific Accucore C18 HPLC column in less than 3 minutes, with UV detection at 220 nm. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore C18, Alprenolol, Atenolol, Beta blockers, HPLC, Metoprolol, Pindolol, Propranolol, SOLA CX, SPE, Urine, UV detection |
| Matrix: | Urine |
For Research Use Only. Not for use in diagnostic procedures.