Find methods for your needs
Quantitative Forensic Analysis of Opiates, Opioids, and Their Metabolites in Human Urine Without Hydrolysis
Description
Analysis of opiate and opioid metabolites in urine is most often done with a hydrolysis step that make total sample preparation time up to 24 hours. The method described here eliminates the hydrolysis step by analyzing the conjugated metabolites intact using a Thermo Scientific Prelude SPLC system for sample preparation and a Thermo Scientific TSQ Endura triple quadrupole mass spectrometer for analysis. For forensic use only.| Market: | Forensics; Toxicology |
| Keywords: | opiates, Metabolites, Forensic Toxicology, Opioids, sample preparation liquid chromatography (SPLC), Mass spectrometry (MS) |
| Matrix: | Urine |
| Author: | Sarah Fair Wandland, Kerry Hassell, Joseph Herman |
| Affiliation: | Thermo Fisher Scientific |