Find methods for your needs
AU206: Improved Separation of Aminobenzamide (2-AB)-Labeled N-glycans from Human α1 Acid-Glycoprotein for Analysis by HPAE-FLD
Description
This application update demonstrates improved chromatographic separation achieved using a Thermo Scientific™ Dionex™ CarboPac™ PA200 column for the HPAE-FLD method described in proposed United States Pharmacopeia (USP) General Chapter <212> to separate 2-AB-labeled N-linked oligosaccharides from human α1 acid-glycoprotein. An ICS-6000 can be used for this application.| Market: | BioPharma |
| Keywords: | CarboPac PA200 column, ICS-5000+, HPAE-PAD, Glycan profiling, AN1130, 2AB-labeling, ICS-6000, TN 20, TN 70671, TN20, TN70671, AU 206, USP Chapter <212> |
| Matrix: | DI water |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA |
For Research Use Only. Not for use in diagnostic procedures.