Find methods for your needs
AN1152: Determination of Carbohydrates in Kombucha Using HPAE-PAD
Description
The method uses the Thermo Scientific™ Dionex™ CarboPac™ PA20 column with electrolytically generated hydroxide eluent and a 15 mil gasket for the gold working electrode to separate and detect glucose, fructose and sucrose in kombucha.| Market: | Food and Beverage |
| Keywords: | beverage, disaccharide, Glucose, Sucrose, Dionex CarboPac PA20 column, fructose, monosaccharide, TN 20, TN 70671, TN20, TN70671, AN 1152 |
| Matrix: | Fermented beverage |
| Author: | Beibei Huang; Jingli Hu; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.