Find methods for your needs
Dionex IonPac AS18-4μm Column Performance Test Using its QAR Method
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report. This record provides an eWorkflow for executing the QAR method on an Integrion HPIC system.| Market: | Environmental |
| Keywords: | Dionex AERS 500 Suppressor, Ion Chromatography (IC), suppressed conductivity detection, Integrion HPIC, Quality Assurance Report (QAR), IonPac AS18-4µm column, Dionex EGC 500 KOH |
| Matrix: | Water |
| Author: | Jingli Hu; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.