Find methods for your needs
AN1155: Determination of Chloride in Infant Formula and Adult Nutritionals
Description
This application note shows an ion chromatography (IC) determination of chloride in 19 SPIFAN (AOAC Stakeholder Panel on Infant Formula and Adult Nutritionals) matrices. The results show that the method fulfills the SPIFAN standard method performance requirements (SMPRs). Repeatability, recovery, linearity, limit of detection, and limit of quantification were determined according to the chloride AOAC SMPR4 and AOAC Single Lab Validation guidelines. An ICS-6000 can be used for this application.| Market: | Food and Beverage |
| Keywords: | AOAC, Dionex IonPac AS15 Column, ICS-5000+, Chloride, Infant Formula, suppressed conductivity detection, SPIFAN, Infant Formula Act of 1980, AOAC Single Lab Validation, standard method performance requirements (SMPR), Ready-to-Feed (RTF) products, Adult Nutritionals, ICS-6000 |
| Matrix: | Adult nutritionals, Infant formula |
| Author: | Manali Aggrawal and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.