Find methods for your needs
AU204: Determination of Inorganic Cations and Ammonium in Environmental Waters Using a Compact Ion Chromatography System
Description
This Application Update revises AN141 using the compact, integrated Dionex Integrion HPIC system with a Dionex IonPac CS16-4μm cation-exchange column. Inorganic cations and ammonium in environmental waters are determined using a 4 μm particle 4 mm i.d. cation-exchange column to shorten analysis time.| Market: | Environmental |
| Keywords: | 4 μm particle column, HPIC, Ammonia, High Ionic strength water, RFIC, wastewater, Drinking water, Integrion, Discharge, IonPac CS16-µm, Soil extract |
| Matrix: | Municipal wastewater |
| Author: | Carl Fisher |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.