Find methods for your needs
Routine, Targeted and Non-Targeted Analysis of Environmental Contaminants of Emerging Concern - Development and Validation of a UHPLC Orbitrap MS Method
Description
Contaminants of emerging concern (CEC) in the environment are generally described as compounds that are unknown or unrecognized, undetected or not routinely monitored, and represent a diverse group of chemicals that may pose a risk to human health and the environment. Due to limited analytical capability and available resources , monitoring of CECs has been focused on selected analytes rather than a holistic approach. We describe a new analytical method that can be used in the quantitative analysis of 61 targeted and 312 non-targeted CECs.| Market: | Environmental |
| Keywords: | UHPLC, Orbitrap MS, Wastewater analysis, Targeted and Untargeted Analysis, Automated Solid Phase Extraction (SPE) |
| Matrix: | Waste water |
| Author: | P. Yang, C. Yang, T. Nguyen, V. Pileggi, K. Akervik, C. Hao, X. Zhao, S. Thach, J. Newman, Y. Lu, S. Kleywegt, S. Tabe, R. Farnood, J. Beck, M. Bromirski and D. Ghosh |
| Affiliation: | Ontario Ministry of the Environment - Canada; University of Toronto - Canada; Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.