Find methods for your needs
Rapid Separation of Eighteen Herbicides Using Advanced UHPLC and Sub-2 µm Solid Core Column Technologies
Description
Environmental monitoring of herbicides used in the production of food crops is becoming more widespread due to their increased use and inevitable leaching from soil into water sources. This application shows the advantages of using the Thermo Scientific Accucore Vanquish C18 1.5 µm UHPLC column and the Vanquish UHPLC system for the separation of 18 herbicides.| Market: | Environmental |
| Keywords: | Herbicides, High resolution, Pesticides, Rapid separation, Vanquish UHPLC, Accucore Vanquish |
| Matrix: | Water |
| Author: | Kristina Kirkham, Luisa Pereira, Derek Hillbeck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.