Find methods for your needs
Determination of Trace Concentrations of Oxyhalides and Bromide in Municipal and Bottled Waters Using a Compact Ion Chromatography System
Description
This record demonstrates that the method in AN167 can be run successfully using an Integrion Ion Chromatography (IC) system. AN 167 shows that IC with a hydroxide-selective Dionex IonPac AS19 column and electrolytic eluent generation is an efficient and sensitive approach for determining trace concentrations of disinfection biproduct (DBP) anions and bromide in municipal and bottled water samples. With this method the analyst can run US EPA Method 300.1 Part B.| Market: | Environmental |
| Keywords: | Dionex IonPac AS19 Column, Oxyhalides, DBP, Water analysis, Integrion |
| Matrix: | Water |
| Author: | Jingli Hu; Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.