Find methods for your needs
IC Assay for Lithium, Sodium, and Calcium in Lithium Carbonate Using a Compact Ion Chromatography System
Description
Lithium carbonate is used to treat a number of mental health problems associated with chemical imbalance in the brain. Sodium and calcium are possible cationic impurities in lithium carbonate preparations. This record shows a method that uses a Thermo Scientific Dionex IonPac CS16 cation-exchange column, an electrolytically generated MSA eluent, and suppressed conductivity detection to determine lithium, sodium, and calcium in lithium carbonate that was published in Application Note 1090. In this record the method is executed using an Integrion ion chromatography system.| Market: | Pharma |
| Keywords: | Drug Substance, IonPac CS16, USP monograph, RFIC, Drug Product |
| Matrix: | Lithium carbonate solution |
| Author: | Carl Fisher |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.