Find methods for your needs
Direct Determination of Existent Chloride and Sulfate in Denatured Ethanol Samples Using a Compact Ion Chromatography System
Description
This method shows the direct determination of existent chloride and sulfate in denatured ethanol samples, previously published in Application Update 194. In this record, the method is executed on an Integrion IC system.| Market: | Chemical |
| Keywords: | biofuel, HPIC, anions, IC, Suppressed Conductivity, IonPac AS22 Column, Integrion, Ion chromatograhy, Denatured alcohol, direct detection, direct determination, AERS 500 carbonate suppressor, potential chloride, Potential sulfate, existent chloride, existent sulfate, Integrion HPIC, Denatured ethanol, ASTM D9317-13 |
| Matrix: | Denatured ethanol |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.