Find methods for your needs
Determination of Anions in Municipal Drinking Water Using a Carbonate Eluents on a Compact Ion Chromatography System
Description
This record shows a 5-min, fast separation of anions in a municipal drinking water sample on the 4µm resin particle format IonPac AS22-Fast-4µm column. This method upgrades Application Brief AB120 by improving the separation using the 4µm resin particle column to produce more efficient peaks. The Integrion ion chromatography system is designed to work with the high pressures generated by the 4µm particles to deliver the improved separation.| Market: | Environmental |
| Keywords: | Fast IC, HPIC, fluoride, Nitrate, IC, Chloride, Sulfate, IonPac AS22 Column, EPA 300.1, Integrion, AERS 500 carbonate suppressor, municipal drinking water, IonPac AS22-Fast-4µm column, carbonate eluents, IonPac AS22-4µm column, APN184 |
| Matrix: | Drinking water |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.