Find methods for your needs
Determination of Perchlorate in Drinking Water Using a Compact Ion Chromatography System
Description
This record shows that AU148 can be run using an Integrion Ion Chromatography. AU148 demonstrates an approved approach for the determination of perchlorate in environmental water samples according to U.S.EPA Method 314.0. This record used an Integrion ion chromatography system with a 4 mm Dionex IonPac AS16 column, eluent generation to produce the KOH eluent, a 1000 μL injection, and suppressed conductivity detection to determine single digit parts per billion concentrations of perchlorate in drinking water.| Market: | Environmental |
| Keywords: | Dionex AERS 500 Suppressor, Dionex IonPac AS16 Column, Water analysis, Integrion, U.S.EPA Method 314.0 |
| Matrix: | Water |
| Author: | Jingli Hu; Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.