Find methods for your needs
Determination of L-Fucose, D-Galactosamine, D-Glucosamine, D-Galactose, D-Glucose and D-Mannose Using the CarboPac PA20 Column QAR Method
Description
A simple HPAE-PAD method for the direct determination of monosaccharides by Reagent Free ICS 5000+ system, requiring only deionized water to electrolytically produce a KOH eluent. The method in this record is the Quality Assurance Report (QAR) method, and therefore can be used to test the performance of a Thermo Scientific Dionex CarboPac PA20 column. An ICS-6000 can be used for this application.Market: | BioPharma; Food and Beverage |
Keywords: | ICS-5000+, Monosaccharides, CarboPac PA20 Column, EGC 500 KOH, ICS-6000, TN 20, TN 70671, TN20, TN70671 |
Matrix: | Water |
Author: | Jingli Hu; Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.